Tisk…

Anti-TNFRSF18 Mouse Monoclonal Antibody [clone: 1E10]

Dodavatel: Abnova
ABNOH00008784-M09EA 10890 CZK
ABNOH00008784-M09
Anti-TNFRSF18 Mouse Monoclonal Antibody [clone: 1E10]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF18.

Type: Primary
Antigen: TNFRSF18
Clonality: Monoclonal
Clone: 1E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR