Tisk…
Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: 1H7]
Dodavatel: Abnova
ABNOMAB1297-M03EA 11580 CZK
ABNOMAB1297-M03
Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: 1H7]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF10A.
Type: Primary
Antigen: TNFRSF10A
Clonality: Monoclonal
Clone: 1H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Objednat