Tisk…

Anti-FAS Mouse Monoclonal Antibody [clone: 7G10]

Dodavatel: Abnova
ABNOMAB1301-M07EA 10890 CZK
ABNOMAB1301-M07
Anti-FAS Mouse Monoclonal Antibody [clone: 7G10]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant FAS.

Type: Primary
Antigen: FAS
Clonality: Monoclonal
Clone: 7G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR