Tisk…

Anti-NGFR Mouse Monoclonal Antibody [clone: 4G5]

Dodavatel: Abnova
ABNOH00004804-M05EA 11230 CZK
ABNOH00004804-M05
Anti-NGFR Mouse Monoclonal Antibody [clone: 4G5]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant NGFR.

Type: Primary
Antigen: NGFR
Clonality: Monoclonal
Clone: 4G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR