Tisk…

Anti-TNFSF18 Mouse Monoclonal Antibody [clone: 4G2]

Dodavatel: Abnova
ABNOH00008995-M15EA 10890 CZK
ABNOH00008995-M15
Anti-TNFSF18 Mouse Monoclonal Antibody [clone: 4G2]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFSF18.

Type: Primary
Antigen: TNFSF18
Clonality: Monoclonal
Clone: 4G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR