Tisk…

Anti-TNFSF18 Mouse Monoclonal Antibody [clone: 3F5]

Dodavatel: Abnova
ABNOH00008995-M17EA 11230 CZK
ABNOH00008995-M17
Anti-TNFSF18 Mouse Monoclonal Antibody [clone: 3F5]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFSF18.

Type: Primary
Antigen: TNFSF18
Clonality: Monoclonal
Clone: 3F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR