Tisk…

Anti-TNFSF18 Mouse Monoclonal Antibody [clone: 2B8]

Dodavatel: Abnova
ABNOH00008995-M14EA 10890 CZK
ABNOH00008995-M14
Anti-TNFSF18 Mouse Monoclonal Antibody [clone: 2B8]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFSF18.

Type: Primary
Antigen: TNFSF18
Clonality: Monoclonal
Clone: 2B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR