Tisk…

Anti-PCIF1 Rabbit Polyclonal Antibody

Dodavatel: Abnova
ABNOPAB29502EA 14770 CZK
ABNOPAB29502
Anti-PCIF1 Rabbit Polyclonal Antibody
Protilátky
PCIF1 polyclonal antibody raised against recombinant human PCIF1.

Type: Primary
Antigen: PCIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR