Tisk…

Anti-NT5E Rabbit Polyclonal Antibody

Dodavatel: Abnova
ABNOPAB29314EA 14770 CZK
ABNOPAB29314
Anti-NT5E Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody raised against recombinant human NT5E.

Type: Primary
Antigen: NT5E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR