Tisk…

Anti-NPPB Rabbit Polyclonal Antibody

Dodavatel: Bioss
BOSSBS-5144REA 9810 CZK
BOSSBS-5144R
Anti-NPPB Rabbit Polyclonal Antibody
Protilátky
This gene is a member of the natriuretic peptide family and encodes a secreted protein which functions as a cardiac hormone. The protein undergoes two cleavage events, one within the cell and a second after secretion into the blood. The protein's biological actions include natriuresis, diuresis, vasorelaxation, inhibition of renin and aldosterone secretion, and a key role in cardiovascular homeostasis. A high concentration of this protein in the bloodstream is indicative of heart failure. Mutations in this gene have been associated with postmenopausal osteoporosis. [provided by RefSeq].

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: NPPB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR