Tisk…

Anti-PCDHA12 Rabbit Polyclonal Antibody

Dodavatel: Bioss
BOSSBS-5825REA 19480 CZK
BOSSBS-5825R
Anti-PCDHA12 Rabbit Polyclonal Antibody
Protilátky
The PCDHA12 gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five, which demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins which may play a role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined.

WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: PCDHA12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR