Tisk…

Anti-CCDC30 Rabbit Polyclonal Antibody

Dodavatel: Bioss
BOSSBS-8097REA 9900 CZK
BOSSBS-8097R
Anti-CCDC30 Rabbit Polyclonal Antibody
Protilátky
Expressed in brain, kidney, pancreas, placenta, liver, thymus and prostate.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CCDC30
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR