Tisk…
Liquid nitrogen transport carrier, CryoPod™
Dodavatel: BioCision
BCISBCS-514EA 122860 CZK
BCISBCS-514
Liquid nitrogen transport carrier, CryoPod™
CryoPod™ Carrier provides safe, reliable and portable <-150 ˚C cryogenic transport for biospecimens for over 4 hours. Use around the laboratory, campus or short transport between facilities. Displays temperature, date and time, and features audible and visual alarms, as well as logging capability.
  • No direct sample contact with LN₂
  • With built-in handle and finger grips
  • Magnetised foam lid for safer transport and insulation
  • Accomodates one standard cryogenic storage box or 2 to 3 small cassettes
It integrates into an (optional) automated filling station or can be manually filled using the included fill kit.
Informace pro objednávání: CryoPod™ Carrier includes green lid and manual fill kit. Optional CryoPod lids available in 4 additional colours.