Tisk…

Anti-HA Mouse Monoclonal Antibody (DyLight® 488) [clone: 16B12]

Dodavatel: Agrisera
AGRIAS152919EA 13200 CZK
AGRIAS152919
Anti-HA Mouse Monoclonal Antibody (DyLight® 488) [clone: 16B12]
Protilátky
Mouse anti-HA is a primary antibody which binds to HA and is directly conjugated toDyLight®488. This is of advantage to shorten assay time by no need to use a secondary antibody to HA. DyLight® is a registered trade mark of Thermofisher Inc., and its subsidaries.,Mouse anti-HA is a primary antibody which binds to HA and is directly conjugated toDyLight®488. This is of advantage to shorten assay time by no need to use a secondary antibody to HA. DyLight® is a registered trade mark of Thermofisher Inc., and its subsidaries.

Type: Primary
Antigen: HA
Clonality: Monoclonal
Clone: 16B12
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR