Tisk…
Human Recombinant Artemin
Dodavatel: Tonbo Biosciences
21-7096-U500EA 47120 CZK
21-7096-U500 21-7096-U020 21-7096-U005 21-7096-U100
Human Recombinant Artemin
Proteiny
Artemin (ARTN) is a secreted disulfide-linked homodimer that belongs to the GDNF family of ligands. It is known to be expressed by embryonic smooth muscle cells and Schwann cells. Artemin promotes sensory neuron survival and likely plays a role in the development of the peripheral nervous system. It signals through the receptor tyrosine kinase (RET)/GFR-alpha 3 receptor complex.