Safety boots, pull-on/zipped, Rock & Roll, Siberian
Supplier: U-Power
Certificates
About this item
Product Information
- Catalog #Supplier #Shoe sizeColourPkAvailabilityPrice
- UPOWRR40754/44RR40754/4444Black1 PAIR1 * 1 PÁR2 880,00 KčSupplier Article Number: RR40754/44Local Article Number: UPOWRR40754/44
Specifications
Environmentally Preferable - UPOWRR40754/45RR40754/4545Black1 PAIR1 * 1 PÁR2 880,00 KčSupplier Article Number: RR40754/45Local Article Number: UPOWRR40754/45
Specifications
Environmentally Preferable - UPOWRR40754/38RR40754/3838Black1 PAIR1 * 1 PÁR2 880,00 KčSupplier Article Number: RR40754/38Local Article Number: UPOWRR40754/38
Specifications
Environmentally Preferable - UPOWRR40754/46RR40754/4646Black1 PAIR1 * 1 PÁR2 880,00 KčSupplier Article Number: RR40754/46Local Article Number: UPOWRR40754/46
Specifications
Environmentally Preferable - UPOWRR40754/41RR40754/4141Black1 PAIR1 * 1 PÁR2 880,00 KčSupplier Article Number: RR40754/41Local Article Number: UPOWRR40754/41
Specifications
Environmentally Preferable - UPOWRR40754/42RR40754/4242Black1 PAIR1 * 1 PÁR2 880,00 KčSupplier Article Number: RR40754/42Local Article Number: UPOWRR40754/42
Specifications
Environmentally Preferable - UPOWRR40754/39RR40754/3939Black1 PAIR1 * 1 PÁR2 880,00 KčSupplier Article Number: RR40754/39Local Article Number: UPOWRR40754/39
Specifications
Environmentally Preferable - UPOWRR40754/40RR40754/4040Black1 PAIR1 * 1 PÁR2 880,00 KčSupplier Article Number: RR40754/40Local Article Number: UPOWRR40754/40
Specifications
Environmentally Preferable - UPOWRR40754/43RR40754/4343Black1 PAIR1 * 1 PÁR2 880,00 KčSupplier Article Number: RR40754/43Local Article Number: UPOWRR40754/43
Specifications
Environmentally Preferable - UPOWRR40754/47RR40754/4747Black1 PAIR1 * 1 PÁR2 880,00 KčSupplier Article Number: RR40754/47Local Article Number: UPOWRR40754/47