Tisk…

Anti-GNL3 Goat Polyclonal Antibody

Dodavatel: Everest Biotechnology
EVBIEB12564EA 7250 CZK
EVBIEB12564
Anti-GNL3 Goat Polyclonal Antibody
Protilátky
Western Blot: Approx 65+75kDa bands observed in nuclear lysates of cell lines HeLa, HepG2 and K562 (calculated MW of 62.0kDa according to NP_055181.3). The observed 75kDa band likely represents the acetylated form. Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: GNL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR