Tisk…
Paladium 5% on activated carbon powder wetted with ca. 50% water, eggshell, unreduced Type A401102-5
Dodavatel: Thermo Scientific
38299.22EA 51370 CZK
38299.22 38299.14 38299.06
Paladium 5% on activated carbon powder wetted with ca. 50% water, eggshell, unreduced Type A401102-5
Palladium
Applications: Aromatic nitro group hydrogenation
Vzorec: Pd
MW: 106,420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03457879
CAS číslo: 7440-05-3