Barium molybdenum oxide ≥99.9% (metals basis excluding Sr)
Dodavatel: Thermo Scientific
|
Varování
|
40205.22EA
6520
CZK
40205.22
Barium molybdenum oxide ≥99.9% (metals basis excluding Sr)
Barium molybdenum oxide
Applications: Electronic and optical equipment, pigment in paints and other protective coatings
Vzorec:
BaMoO₄ MW: 297,28 g/mol Bod tání: ∼ 1600 °CC Hustota: 4,98 Storage Temperature: Ambient |
MDL Number:
MFCD00014182 CAS číslo: 7787-37-3 EINECS: 232-111-7 UN: 1564 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...