Barium-2-methoxyethoxide 10% (w/v) in 2-methoxyethanol , 99.5% (metals basis)
Dodavatel: Thermo Scientific
41256.ACEA
4090
CZK
41256.AC
Barium-2-methoxyethoxide 10% (w/v) in 2-methoxyethanol , 99.5% (metals basis)
Barium 2-methoxyethoxide
Vzorec:
C₆H₁₄BaO₄ |
MDL Number:
MFCD00673672 CAS číslo: 115503-13-4 UN: 3274 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...