Oxid hlinitý, Drysphere® desiccant, with indicator 20%
Synonyms:
Alumina C
46126.30EA
1420
CZK
46126.30
46126.A1
Oxid hlinitý, Drysphere® desiccant, with indicator 20%
Oxid hlinitý
Vzorec:
Al₂O₃ MW: 101,96 g/mol Bod varu: 2980 °C Bod tání: 2045 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS číslo: 1344-28-1 EINECS: 215-691-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...