Tisk…

Nickel aluminium alloy according to Raney, non-activated

Dodavatel: Thermo Scientific
Nebezpečí

87676.36EA 6040 CZK
87676.36 87676.A3 87676.18
Nickel aluminium alloy according to Raney, non-activated
Nickel aluminium alloy according to Raney

Nickel Aluminum is used for the manufacture of aluminum-nickel alloy powder, and it is used in the chemical, pharmaceutical.

Vzorec: Ni:Al;₅₀:₅₀%
Storage Temperature: Ambient
MDL Number: MFCD00167392
CAS číslo: 12635-29-9
UN: 3208
ADR: 4.3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR