Tisk…

Paladium 0,5% on alumina, pellets 3.18 mm (0.125 in), unreduced

Dodavatel: Thermo Scientific

89114.36EA 28730 CZK
89114.36 89114.14 89114.22
Paladium 0,5% on alumina, pellets 3.18 mm (0.125 in), unreduced
Palladium
Vzorec: Pd
MW: 106,420g,mol
Storage Temperature: Ambient
MDL Number: MFCD03457879
CAS číslo: 7440-05-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR