(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid 97%
Dodavatel: Thermo Scientific
H26919.03EA
2180
CZK
H26919.03
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid 97%
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid
Vzorec:
C₈H₁₆N₂O₄ MW: 204,23 g/mol Bod tání: ∼ 210 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00236843 CAS číslo: 73259-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...