5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid 95%
Dodavatel: Thermo Scientific
|
Varování
|
H32316.03EA
6080
CZK
H32316.03
5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid 95%
5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid
Vzorec:
C₇H₃BrF₃NO₂ MW: 270,01 g/mol Bod tání: 235…238 °C Storage Temperature: Ambient |
MDL Number:
MFCD10687236 CAS číslo: 749875-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...