4-Methylthiazole-5-carboxylic acid 97%
|
Varování
|
H32662.09EA
5670
CZK
H32662.09
4-Methylthiazole-5-carboxylic acid 97%
4-Methylthiazole-5-carboxylic acid
Vzorec:
C₅H₅NO₂S MW: 143,17 g/mol Bod tání: 287 °C Storage Temperature: Ambient |
MDL Number:
MFCD00626872 CAS číslo: 20485-41-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...