Ethyl-2-fluoroisobutyrate 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H32931.06EA
8310
CZK
H32931.06
Ethyl-2-fluoroisobutyrate 97%
Ethyl-2-fluoroisobutyrate
Vzorec:
C₆H₁₁FO₂ MW: 134,15 g/mol Bod varu: 122…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD08062432 CAS číslo: 55816-69-8 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...