Tisk…

3-Methyl-5-(trifluoromethyl)benzaldehyde 97%

Varování

H33598.03EA 2300 CZK
H33598.03 H33598.06
3-Methyl-5-(trifluoromethyl)benzaldehyde 97%
3-Methyl-5-(trifluoromethyl)benzaldehyde
Vzorec: C₉H₇F₃O
MW: 188,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652409
CAS číslo: 116070-39-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR