1-(Pyrimidin-2-yl)piperidine-4-carboxylic acid 95%
|
Varování
|
H50351.03EA
7750
CZK
H50351.03
1-(Pyrimidin-2-yl)piperidine-4-carboxylic acid 95%
1-(Pyrimidin-2-yl)piperidine-4-carboxylic acid
Vzorec:
C₁₀H₁₃N₃O₂ MW: 207,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00664474 CAS číslo: 303144-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...