(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid 95%
H52175.03EA
10890
CZK
H52175.03
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid 95%
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid
Vzorec:
C₁₁H₁₉NO₄ MW: 229,3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01862939 CAS číslo: 56502-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...