Tisk…

2,6-Dichloro-4-methylnicotinic acid 98%

Varování

H52347.06EA 19290 CZK
H52347.06 H52347.03 H52347.MD
2,6-Dichloro-4-methylnicotinic acid 98%
2,6-Dichloro-4-methylnicotinic acid
Vzorec: C₇H₅Cl₂NO₂
MW: 206,03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02932724
CAS číslo: 62774-90-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR