Tisk…

4-Boc-1-(6-bromo-2-pyridyl)piperazine 97%

Nebezpečí

H55209.14EA 19010 CZK
H55209.14
4-Boc-1-(6-bromo-2-pyridyl)piperazine 97%
4-Boc-1-(6-bromo-2-pyridyl)piperazine
Vzorec: C₁₄H₂₀BrN₃O₂
MW: 342,237 g/mol
Bod tání: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD07369773
CAS číslo: 331767-56-7
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR