Tisk…

(S)-2-Aminobutanamide hydrochloride 99%

Dodavatel: Thermo Scientific
Varování

H55576.06EA 7460 CZK
H55576.06 H55576.MD H55576.03
(S)-2-Aminobutanamide hydrochloride 99%
(S)-2-Aminobutanamide hydrochloride
Vzorec: C₄H₁₁ClN₂O
MW: 138,60 g/mol
Bod tání: 259…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00136565
CAS číslo: 7682-20-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR