Tisk…

Bis(4-fluorobenzyl)amine 97%

Dodavatel: Thermo Scientific
Nebezpečí

H56467.03EA 15240 CZK
H56467.03 H56467.MD
Bis(4-fluorobenzyl)amine 97%
Bis(4-fluorobenzyl)amine
Vzorec: C₁₄H₁₃F₂N
Storage Temperature: Ambient
MDL Number: MFCD05155916
CAS číslo: 134227-41-1
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR