Tisk…

Methyl-3-amino-6-iodopyrazine-2-carboxylate 96%

Dodavatel: Thermo Scientific
Varování

H63026.03EA 6080 CZK
H63026.03 H63026.06 H63026.MD
Methyl-3-amino-6-iodopyrazine-2-carboxylate 96%
Methyl-3-amino-6-iodopyrazine-2-carboxylate
Vzorec: C₆H₆IN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09909640
CAS číslo: 1458-16-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR