Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate 95%
Dodavatel: Thermo Scientific
H63751.MDEA
2870
CZK
H63751.MD
Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate 95%
Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate
Vzorec:
C₁₀H₁₃NO₃ MW: 195,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00030352 CAS číslo: 2199-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...