4-Amino-5,6-dichloropyrimidine 98%
|
Varování
|
H64691.06EA
4670
CZK
H64691.06
4-Amino-5,6-dichloropyrimidine 98%
4-Amino-5,6-dichloropyrimidine
Vzorec:
C₄H₃Cl₂N₃ MW: 163,99 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09837293 CAS číslo: 310400-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...