Tisk…

5-Bromo-1H-indole-3-carboxylic acid 97%

Dodavatel: Thermo Scientific
Varování

H64834.03EA 3810 CZK
H64834.03 H64834.06 H64834.MD
5-Bromo-1H-indole-3-carboxylic acid 97%
5-Bromo-1H-indole-3-carboxylic acid
Vzorec: C₉H₆BrNO₂
MDL Number: MFCD05664007
CAS číslo: 10406-06-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR