1-(1-Boc-piperidino)-4-bromo-1H-pyrazole 98%
Dodavatel: Thermo Scientific
|
Varování
|
H65315.03EA
3090
CZK
H65315.03
H65315.06
1-(1-Boc-piperidino)-4-bromo-1H-pyrazole 98%
1-(1-Boc-piperidino)-4-bromo-1H-pyrazole
Vzorec:
C₁₃H₂₀BrN₃O₂ MW: 330,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09831980 CAS číslo: 877399-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...