Tisk…

Human angiotensin 2

Dodavatel: Thermo Fisher Scientific
J60866.MBEA 11230 CZK
J60866.MB
Human angiotensin 2
Proteiny
CAS No.: 4474-91-3
Molecular Formula: C50H71N13O12
Formula Weight: 1046.19
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00133092
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR