Tisk…
TEA, 0,2 M buffer soln., pH 7,0
Dodavatel: Thermo Fisher Scientific
J61546.AKEA 1430 CZK
ALFAJ61546.AK ALFAJ61546.AP
TEA, 0,2 M buffer soln., pH 7,0
CAS No.: 102-71-6
Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00002855