Tisk…
SOB Medium
Dodavatel: Thermo Fisher Scientific
J62210.AKEA 3280 CZK
J62210.AK
SOB Medium
Média mikrobiologická Připravená mikrobiologická média
SOB Medium was developed by Hanahan as a nutritionally rich growth medium for preparation and transformation of competent cells. Growth of E. coli in SOB medium results in higher transformation efficiencies of plasmids.
  • Yellow-brown Liquid
Contains 20 g/l tryptone, 5g/l yeast extract, 2,4 g/l MgSO4, 0,5 g/l NaCl, and 0,186 g/l KCl.