Tisk…
Ficoll® based agarose gel loading dyes
Dodavatel: Thermo Fisher Scientific
J62800.ACEA 2890 CZK
ALFAJ62800.AC ALFAJ62800.AB
Ficoll® based agarose gel loading dyes
Ficoll® based agarose gel loading dyes.
Informace pro objednávání: Contains bromophenol blue (0,25 %), xylene cyanol FF (0,25 %) and Ficoll® (15 %) in water.