Tisk…

BLOTTO in TBS

Dodavatel: Thermo Fisher Scientific
J63238.AKEA 1560 CZK
ALFAJ63238.AK
BLOTTO in TBS
Storage Temperature: −30 to −10 °C
Physical Form: Liquid
MDL No.: MFCD00163095

Applications: Blocks non-specific binding site for antibodies on the membrane
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR