Tisk…

Bovine mucin

Dodavatel: Thermo Fisher Scientific
J63859.MCEA 0 CZK
ALFAJ63859.MC
Bovine mucin
CAS No.: 84195-52-8
Storage Temperature: 2 to 8 °C
Physical Form: Powder, 400 to 4000 kDa
Appearance: Pale brown
MDL No.: MFCD00131629

Applications: Neuraminidase substrate
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR