Tisk…

Sirtinol

Dodavatel: Thermo Scientific
Varování

J64486.MAEA 5600 CZK
J64486.MA
Sirtinol
Sirtinol

Applications: Sirtinol is a cell permeable specific inhibitor of sirtuin NAD-dependant deacetylases

Vzorec: C₂₆H₂₂N₂O₂
MW: 394,46 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00810186
CAS číslo: 410536-97-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR