Tisk…

Cathepsin B inhibitor 4

Dodavatel: Alfa Aesar
J64737.MAEA 17990 CZK
J64737.MA
Cathepsin B inhibitor 4
Proteiny
CAS No.: 147859-80-1
Molecular Formula: C19H31N3O6
Formula Weight: 397,46
Storage Temperature: −30 to −10 °C
Physical Form: Powder
Appearance: White
Solubility: Soluble in DMSO
MDL No.: MFCD03452890

Applications: A cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR