Tisk…
Reagent reservoirs, Pierce™ ELISA
Dodavatel: Thermo Scientific
PIER15075EA 0 CZK
PIER15075
Reagent reservoirs, Pierce™ ELISA
Disposable reagent reservoirs molded from high-impact PS. Reservoirs facilitate repeated multi-channel pipetting for reagent delivery to microplates.
  • Sterile and durable
  • Reservoirs facilitate repeated pick-up of liquid reagents by multi-channel pipettors for delivery to 96-well plates