Tisk…
Classic magnetic IP/Co-IP kit, Pierce™
Dodavatel: Thermo Fisher Scientific
Pierce
PIER88804EA 19370 CZK
PIER88804
Classic magnetic IP/Co-IP kit, Pierce™
Systémy pro purifikaci proteinů Protein Purification Kits
The Pierce™ Classic Magnetic IP/Co-IP Kit enables highly effective and efficient immunoprecipitation (IP) and co-immunoprecipitation (co-IP) of antigens and protein-complexes using less than 10 µg of antibody and a magnetic separator.
  • No leaching of protein A/G in the presence of detergents, low pH buffers or common mass spectrometry solvents
  • Immunoprecipitate in as little as 30 minutes to reduce background and improve the capture of transient protein complexes
  • Efficient and convenient with all essential buffers to complete immunoprecipitation experiments
  • Magnetic beads based on Protein A/G recombinant protein ensures compatibility with most primary antibodies
Informace o dodávce: The kit contains Protein A/G magnetic beads, 1 ml; IP lysis/wash buffer, 2×50 ml; lane marker sample buffer (5X), 5 ml; elution buffer, 5 ml; and neutralisation buffer, 0,5 ml.