Tisk…
Transfer buffer, 10X Western Blot Transfer buffer, Pierce™
Dodavatel: Thermo Scientific
PIER35040EA 4290 CZK
PIER35040
Transfer buffer, 10X Western Blot Transfer buffer, Pierce™
This transfer buffer is a methanol-free, space saving, stock solution for the preparation of methanol-free transfer buffer that is used with cooled, tank (wet) Western blotting transfer apparatus.
  • Easy to use - no packets to open, no powder to dissolve, and no methanol required
  • Increased accuracy
  • Saves time and space
For research use only. Not for use in diagnostic procedures.